Assay Details
| Gene Symbol: foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142162.1 | NP_001135634.1 | 8 - 9 | 1894 | 62 | |
| GenBank mRNA | BC168486.1 | - | 8 - 9 | 1894 | 62 | |
More Information
Set Membership: |
|

foxk2
Genomic Map