Assay Details
| Gene Symbol: fdft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142170.3 | NP_001135642.3 | 1 - 2 | 108 | 61 | |
| GenBank mRNA | BC168496.1 | - | 1 - 2 | 108 | 61 | |
More Information
Set Membership: |
|

fdft1
Genomic Map