Assay Details
| Gene Symbol: chpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142222.1 | NP_001135694.1 | 3 - 4 | 539 | 63 | |
| XM_012958717.2 | XP_012814171.1 | 3 - 4 | 657 | 63 | ||
| XM_018091982.1 | XP_017947471.1 | 3 - 4 | 657 | 63 | ||
| GenBank mRNA | BC168802.1 | - | 3 - 4 | 539 | 63 | |
| CT030057.1 | - | 3 - 4 | 612 | 63 | ||
More Information
Set Membership: |
|

chpt1
Genomic Map