Assay Details
| Gene Symbol: ribc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142228.2 | NP_001135700.1 | 3 - 4 | 549 | 79 | |
| XM_012968121.2 | XP_012823575.1 | 4 - 5 | 645 | 79 | ||
| XM_012968122.2 | XP_012823576.1 | 4 - 5 | 646 | 79 | ||
| XM_012968123.1 | XP_012823577.1 | 4 - 5 | 647 | 79 | ||
| XM_012968124.2 | XP_012823578.1 | 4 - 5 | 568 | 79 | ||
| GenBank mRNA | BC168813.1 | - | 3 - 4 | 551 | 79 | |
| CR761888.2 | - | 4 - 5 | 669 | 79 | ||
More Information
Set Membership: |
|

ribc1
Genomic Map