Assay Details
| Gene Symbol: mrps11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142250.1 | NP_001135722.1 | 4 - 5 | 439 | 59 | |
| GenBank mRNA | BC169180.1 | - | 4 - 5 | 439 | 59 | |
| CU075403.1 | - | 4 - 5 | 427 | 59 | ||
More Information
Set Membership: |
|

mrps11
Genomic Map