Assay Details
| Gene Symbol: fgf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142822.1 | NP_001136294.1 | 2 - 3 | 396 | 89 | |
| GenBank mRNA | FJ428266.1 | - | 2 - 3 | 396 | 89 | |
More Information
Set Membership: |
|

fgf4
Genomic Map