Assay Details
| Gene Symbol: fgf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142825.1 | NP_001136297.1 | 1 - 2 | 232 | 61 | |
| GenBank mRNA | FJ430563.1 | - | 1 - 2 | 232 | 61 | |
More Information
Set Membership: |
|

fgf19
Genomic Map