Assay Details
| Gene Symbol: fgf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143927.1 | NP_001137399.1 | 1 - 2 | 364 | 63 | |
| GenBank mRNA | FJ474092.1 | - | 1 - 2 | 364 | 63 | |
More Information
Set Membership: |
|

fgf20
Genomic Map