Assay Details
| Gene Symbol: henmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159540.1 | NP_001153012.1 | 3 - 4 | 334 | 65 | |
| XM_012960696.2 | XP_012816150.1 | 4 - 5 | 665 | 65 | ||
| XM_012960697.2 | XP_012816151.1 | 4 - 5 | 519 | 65 | ||
| XM_012960698.2 | XP_012816152.1 | 4 - 5 | 451 | 65 | ||
| XM_012960699.1 | XP_012816153.1 | 3 - 4 | 399 | 65 | ||
| GenBank mRNA | EU338244.1 | - | 3 - 4 | 334 | 65 | |
More Information
Set Membership: |
|

henmt1
Genomic Map