Assay Details
| Gene Symbol: c6orf120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004942.1 | NP_001004942.1 | 2 | 550 | 76 | |
| XM_012962884.2 | XP_012818338.1 | 2 | 609 | 76 | ||
| XM_012962885.2 | XP_012818339.1 | 2 | 799 | 76 | ||
| XM_018093840.1 | XP_017949329.1 | 2 | 582 | 76 | ||
| GenBank mRNA | BC075425.1 | - | 2 | 550 | 76 | |
More Information
Set Membership: |
|

c6orf120
Genomic Map