Assay Details
| Gene Symbol: isg20l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004997.1 | NP_001004997.1 | 2 | 1051 | 97 | |
| XM_012968236.2 | XP_012823690.1 | 3 | 1128 | 97 | ||
| XM_012968237.2 | XP_012823691.1 | 3 | 1127 | 97 | ||
| GenBank mRNA | BC075592.1 | - | 2 | 1051 | 97 | |
| CU075342.1 | - | 2 | 1050 | 97 | ||
More Information
Set Membership: |
|

isg20l2
Genomic Map