Assay Details
| Gene Symbol: klk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011435.2 | NP_001011435.2 | 2 | 159 | 102 | |
| XM_018095375.1 | XP_017950864.1 | 2 | 160 | 102 | ||
| GenBank mRNA | BC088079.1 | - | 2 | 153 | 102 | |
| EL763762.1 | - | 2 | 159 | 102 | ||
More Information
Set Membership: |
|

klk15
Genomic Map