Assay Details
| Gene Symbol: pqlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015930.3 | NP_001015930.2 | 7 | 1813 | 103 | |
| GenBank mRNA | BC121512.1 | - | 7 | 1728 | 103 | |
| BC154862.1 | - | 7 | 1795 | 103 | ||
| CR926378.2 | - | 4 | 1645 | 103 | ||
More Information
Set Membership: |
|

pqlc3
Genomic Map