Assay Details
Gene Symbol: tcf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016506.2 | NP_001016506.1 | 451 | 83 | ||
GenBank mRNA | BC171047.1 | - | 444 | 83 | ||
BC171049.1 | - | 444 | 83 | |||
CR760785.2 | - | 451 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
