Assay Details
| Gene Symbol: mafb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030523.1 | NP_001025694.1 | 1 | 548 | 95 | |
| GenBank mRNA | BC094951.1 | - | 1 | 548 | 95 | |
| DQ018731.1 | - | 1 | 278 | 95 | ||
More Information
Set Membership: |
|

mafb
Genomic Map