Assay Details
| Gene Symbol: fzd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097391.3 | NP_001090860.2 | 1 | 1548 | 108 | |
| GenBank mRNA | AL959610.2 | - | 1 | 147 | 108 | |
| BC135224.1 | - | 1 | 1555 | 108 | ||
| DT402720.1 | - | 1 | 119 | 108 | ||
More Information
Set Membership: |
|

fzd8
Genomic Map