Assay Details
| Gene Symbol: dnajc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102878.1 | NP_001096348.1 | 2 | 1646 | 106 | |
| GenBank mRNA | BC135818.1 | - | 2 | 1646 | 106 | |
More Information
Set Membership: |
|

dnajc18
Genomic Map