Assay Details
| Gene Symbol: dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102997.1 | NP_001096467.1 | 856 | 104 | ||
| GenBank mRNA | BC136203.1 | - | 856 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dctn3| Gene Symbol: dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102997.1 | NP_001096467.1 | 856 | 104 | ||
| GenBank mRNA | BC136203.1 | - | 856 | 104 | ||
Set Membership: |
|