Assay Details
| Gene Symbol: gnrhr2/nmi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114076.1 | NP_001107548.1 | 3 | 854 | 107 | |
| XM_018091928.1 | XP_017947417.1 | 5 | 1189 | 107 | ||
| GenBank mRNA | EF166087.1 | - | 3 | 854 | 107 | |
More Information
Set Membership: |
|

gnrhr2/nmi
Genomic Map