Assay Details
| Gene Symbol: ocm.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114201.1 | NP_001107673.1 | 5 | 540 | 110 | |
| GenBank mRNA | BC084465.1 | - | 4 | 470 | 109 | |
| BC153701.1 | - | 5 | 540 | 110 | ||
| CU464108.1 | - | 5 | 532 | 109 | ||
More Information
Set Membership: |
|

ocm.2
Genomic Map