Assay Details
| Gene Symbol: yipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126994.1 | NP_001120466.1 | 947 | 84 | ||
| GenBank mRNA | BC161293.1 | - | 947 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

yipf3| Gene Symbol: yipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126994.1 | NP_001120466.1 | 947 | 84 | ||
| GenBank mRNA | BC161293.1 | - | 947 | 84 | ||
Set Membership: |
|