Assay Details
| Gene Symbol: Atg4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144017.1 | NP_001137489.1 | 1350 | 64 | ||
| GenBank mRNA | BT039657.1 | - | 1280 | 64 | ||
| EU976029.1 | - | 1350 | 64 | |||
| FJ445000.1 | - | 1140 | 64 | |||
| FJ445001.1 | - | 1083 | 64 | |||
| FJ445002.1 | - | 1173 | 64 | |||
| FJ445003.1 | - | 1055 | 64 | |||
| FJ445004.1 | - | 925 | 64 | |||
More Information
Set Membership: |
|

Atg4b
Genomic Map