Assay Details
| Gene Symbol: IDP2552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153102.1 | NP_001146574.1 | 1219 | 79 | ||
| GenBank mRNA | BT055704.1 | - | 1219 | 79 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IDP2552| Gene Symbol: IDP2552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153102.1 | NP_001146574.1 | 1219 | 79 | ||
| GenBank mRNA | BT055704.1 | - | 1219 | 79 | ||