Assay Details
| Gene Symbol: LOC100283435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156336.1 | NP_001149808.1 | 368 | 51 | ||
| GenBank mRNA | EU964750.1 | - | 368 | 51 | ||
More Information
Set Membership: |
|

LOC100283435
Genomic Map