Assay Details
| Gene Symbol: LOC100284118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157015.1 | NP_001150487.1 | 347 | 85 | ||
| GenBank mRNA | BT038766.1 | - | 347 | 85 | ||
| BT054479.1 | - | 256 | 85 | |||
| BT060497.1 | - | 310 | 85 | |||
| BT064091.1 | - | 358 | 85 | |||
| EU967137.1 | - | 456 | 85 | |||
More Information
Set Membership: |
|

LOC100284118
Genomic Map