Assay Details
| Gene Symbol: AC207628.4_FG005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155636.1 | NP_001149108.1 | 658 | 75 | ||
| GenBank mRNA | EU962114.1 | - | 658 | 75 | ||
More Information
Set Membership: |
|

AC207628.4_FG005
Genomic Map