Assay Details
| Gene Symbol: LOC100285435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158328.1 | NP_001151800.1 | 456 | 83 | ||
| GenBank mRNA | EU972232.1 | - | 456 | 83 | ||
More Information
Set Membership: |
|

LOC100285435
Genomic Map