Assay Details
| Gene Symbol: LOC100273587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148004.1 | NP_001141476.1 | 371 | 54 | ||
| GenBank mRNA | BT041443.1 | - | 371 | 54 | ||
| BT063196.1 | - | 268 | 54 | |||
| EU960388.1 | - | 356 | 54 | |||
More Information
Set Membership: |
|

LOC100273587
Genomic Map