Assay Details
| Gene Symbol: LOC100282620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155528.1 | NP_001149000.1 | 353 | 83 | ||
| GenBank mRNA | EU961643.1 | - | 353 | 83 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100282620| Gene Symbol: LOC100282620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155528.1 | NP_001149000.1 | 353 | 83 | ||
| GenBank mRNA | EU961643.1 | - | 353 | 83 | ||