Assay Details
Gene Symbol: LOC100285960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158849.1 | NP_001152321.1 | 930 | 75 | ||
GenBank mRNA | EU974893.1 | - | 930 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
