Assay Details
| Gene Symbol: LOC100282579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155487.1 | NP_001148959.1 | 492 | 91 | ||
| GenBank mRNA | EU961527.1 | - | 492 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100282579| Gene Symbol: LOC100282579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155487.1 | NP_001148959.1 | 492 | 91 | ||
| GenBank mRNA | EU961527.1 | - | 492 | 91 | ||