Assay Details
| Gene Symbol: LOC100283075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155977.1 | NP_001149449.1 | 469 | 100 | ||
| GenBank mRNA | EU963368.1 | - | 469 | 100 | ||
More Information
Set Membership: |
|

LOC100283075
Genomic Map