Assay Details
| Gene Symbol: lox9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112507.1 | NP_001105977.1 | 1869 | 61 | ||
| GenBank mRNA | DQ335767.1 | - | 1869 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lox9| Gene Symbol: lox9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112507.1 | NP_001105977.1 | 1869 | 61 | ||
| GenBank mRNA | DQ335767.1 | - | 1869 | 61 | ||
Set Membership: |
|