Assay Details
| Gene Symbol: LOC100279948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001152898.1 | NP_001146370.1 | 1074 | 73 | ||
| GenBank mRNA | BT055196.1 | - | 1074 | 73 | ||
More Information
Set Membership: |
|

LOC100279948
Genomic Map