Assay Details
| Gene Symbol: LOC100282174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155086.1 | NP_001148558.1 | 649 | 76 | ||
| GenBank mRNA | EU959922.1 | - | 649 | 76 | ||
More Information
Set Membership: |
|

LOC100282174
Genomic Map