Assay Details
Gene Symbol: LOC100283552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156452.1 | NP_001149924.1 | 1031 | 96 | ||
GenBank mRNA | EU965099.1 | - | 1031 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
