Assay Details
| Gene Symbol: LOC100286050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158938.1 | NP_001152410.1 | 258 | 90 | ||
| GenBank mRNA | EU975346.1 | - | 258 | 90 | ||
More Information
Set Membership: |
|

LOC100286050
Genomic Map