Assay Details
| Gene Symbol: rpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148634.1 | NP_001142106.1 | 578 | 74 | ||
| GenBank mRNA | BT042645.1 | - | 578 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpl19| Gene Symbol: rpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148634.1 | NP_001142106.1 | 578 | 74 | ||
| GenBank mRNA | BT042645.1 | - | 578 | 74 | ||
Set Membership: |
|