Assay Details
| Gene Symbol: bas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156350.1 | NP_001149822.1 | 584 | 64 | ||
| GenBank mRNA | BT040236.1 | - | 466 | 64 | ||
| EU964791.1 | - | 584 | 64 | |||
More Information
Set Membership: |
|

bas1
Genomic Map