Assay Details
| Gene Symbol: LOC100272577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147042.1 | NP_001140514.1 | 406 | 77 | ||
| GenBank mRNA | BT038979.1 | - | 406 | 77 | ||
| EU956305.1 | - | 429 | 77 | |||
More Information
Set Membership: |
|

LOC100272577
Genomic Map