Assay Details
| Gene Symbol: LOC100272577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147042.1 | NP_001140514.1 | 761 | 86 | ||
| GenBank mRNA | BT038979.1 | - | 761 | 86 | ||
| EU956305.1 | - | 784 | 86 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100272577| Gene Symbol: LOC100272577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147042.1 | NP_001140514.1 | 761 | 86 | ||
| GenBank mRNA | BT038979.1 | - | 761 | 86 | ||
| EU956305.1 | - | 784 | 86 | |||