Assay Details
| Gene Symbol: LOC100282677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155585.1 | NP_001149057.1 | 1051 | 57 | ||
| GenBank mRNA | EU961896.1 | - | 1051 | 57 | ||
More Information
Set Membership: |
|

LOC100282677
Genomic Map