Assay Details
| Gene Symbol: pco102443(516) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158518.1 | NP_001151990.1 | 470 | 76 | ||
| GenBank mRNA | EU965494.1 | - | 199 | 76 | ||
| EU973167.1 | - | 470 | 76 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

pco102443(516)| Gene Symbol: pco102443(516) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158518.1 | NP_001151990.1 | 470 | 76 | ||
| GenBank mRNA | EU965494.1 | - | 199 | 76 | ||
| EU973167.1 | - | 470 | 76 | |||