Assay Details
| Gene Symbol: LOC100285126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158021.1 | NP_001151493.1 | 338 | 71 | ||
| GenBank mRNA | BT070051.1 | - | 301 | 71 | ||
| EU970959.1 | - | 338 | 71 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100285126| Gene Symbol: LOC100285126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158021.1 | NP_001151493.1 | 338 | 71 | ||
| GenBank mRNA | BT070051.1 | - | 301 | 71 | ||
| EU970959.1 | - | 338 | 71 | |||