Assay Details
| Gene Symbol: LOC100285126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158021.1 | NP_001151493.1 | 656 | 62 | ||
| GenBank mRNA | BT070051.1 | - | 619 | 62 | ||
| EU970959.1 | - | 656 | 62 | |||
More Information
Set Membership: |
|

LOC100285126
Genomic Map