Assay Details
| Gene Symbol: LOC100285551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158442.1 | NP_001151914.1 | 675 | 65 | ||
| GenBank mRNA | EU972823.1 | - | 675 | 65 | ||
More Information
Set Membership: |
|

LOC100285551
Genomic Map