Assay Details
| Gene Symbol: LOC100285685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158576.1 | NP_001152048.1 | 1182 | 64 | ||
| GenBank mRNA | EU973422.1 | - | 1182 | 64 | ||
More Information
Set Membership: |
|

LOC100285685
Genomic Map