Assay Details
Gene Symbol: gpm517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001152483.1 | NP_001145955.1 | 545 | 81 | ||
GenBank mRNA | BT054321.1 | - | 545 | 81 | ||
BT061586.1 | - | 673 | 81 |
Gene Symbol: LOC100501645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001196325.1 | NP_001183254.1 | 710 | 81 | ||
GenBank mRNA | BT085858.1 | - | 710 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT085858.1, NM_001196325.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
