Assay Details
| Gene Symbol: gpm517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001152483.1 | NP_001145955.1 | 953 | 85 | ||
| GenBank mRNA | BT054321.1 | - | 953 | 85 | ||
| BT061586.1 | - | 1193 | 85 | |||
More Information
Set Membership: |
|

gpm517
Genomic Map