Assay Details
| Gene Symbol: iso3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111728.1 | NP_001105198.1 | 2251 | 71 | ||
| GenBank mRNA | AY172634.2 | - | 2251 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

iso3| Gene Symbol: iso3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111728.1 | NP_001105198.1 | 2251 | 71 | ||
| GenBank mRNA | AY172634.2 | - | 2251 | 71 | ||
Set Membership: |
|